Cap-It Tool (for domed cap strips)
Thermo Scientific™
Cap-It Tool (for domed cap strips)
The Thermo Scientific Cap-It Tool makes it fast and efficient to add domed caps to tube strips and plates andRead more
Have Questions?
Catalog NumberQuantity
AB05361 unit
Catalog number AB0536
Price (USD)
41.06
Each
Quantity:
1 unit
Price (USD)
41.06
Each
The Thermo Scientific Cap-It Tool makes it fast and efficient to add domed caps to tube strips and plates and helps seal multiple tubes at the same time.
Features of the Thermo Scientific Cap-It Tool
• Designed to assist in applying domed caps to tube strips and plates
• Minimizes the pressure required to seal the caps effectively
• Enables multiple tubes to be sealed simultaneously
Related Products
Adhesive Seal Applicator
Specifications
Compatibility8-Well strips, 96-Well Plates, 96-Well Plate, Fast (0.1 ml), 48-Well Plates, 24-Well Plates
Quantity1 unit
Unit SizeEach