Adhesive PCR Plate Seals
Thermo Scientific™
Adhesive PCR Plate Seals
Thermo Scientific Adhesive PCR Plate Seals are a reliable method of sealing plates for PCR applications. The high integrity sealingRead more
Have Questions?
Catalog NumberQuantity
AB0558100 sheets
Catalog number AB0558
Price (USD)
191.65
Online exclusive
226.00
Save 34.35
Each
Quantity:
100 sheets
Price (USD)
191.65
Online exclusive
226.00
Save 34.35
Each
Thermo Scientific Adhesive PCR Plate Seals are a reliable method of sealing plates for PCR applications. The high integrity sealing materials give the best protection against evaporation for high efficiency PCR.
Features of Thermo Scientific Adhesive PCR Plate Seals
  • Clear for sample inspection
  • Strong adhesive prevents evaporation during thermal cycling or sample storage
  • No sticky residue on the plate when the seal is peeled off
  • End tabs for easy seal application and removal
  • Recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
  • Temperature range: -20°C to 120°C
Specifications
DescriptionPeelable adhesive PCR film
No. per Pack100
Product TypeFilm, Seal, or Applicator
Quantity100 sheets
Temperature Range (Metric)-20°C to +120°C
AdhesiveAdhesive Clear Seal
ColorClear
Plate Compatibility96/384-well Plate
Unit SizeEach
Frequently asked questions (FAQs)
Are Thermo Scientific adhesive seals free of DNase, RNase, and human DNA?
Whilst Thermo Scientific adhesive seals are manufactured in a clean facility, they are not certified free of DNase, RNase, and human DNA.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
I am having problems with evaporation when using an adhesive seal. What do you recommend?
To prevent evaporation when using adhesive seals, firm application pressure must be applied to establish robust adhesive contact and bond strength around well rims. Adhesive seals such as ABsolute qPCR Seal (Cat. No. AB-1170) require sufficient pressure due to their non-tacky, pressure-sensitive adhesive. Adhesive Seal Applicator (Cat. No. AB-1391) or a similar tool may be used for optimal application of the seals. Alternatively, we recommend Thermo Scientific VersiCap Mats which show low sample evaporation with the performance of a cap. VersiCap Mats (Cat. Nos. AB-1810, AB-1815, AB-1820) offer comparable convenience and are easier to apply uniformly than adhesive seals.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What is the shelf life of the Adhesive PCR Plate Seals (Cat. No. AB0558)?
Adhesive PCR Plate Seals are covered under our general 1-year warranty and are guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.