ABgene Storage Plate, 96-well, 1.2 mL, individually wrapped, black

Catalog number: AB0564K

Thermo Scientific™  Related applications: Cell Counting, Viability, & Cryopreservation | Microplates, Dishes and Flasks

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific ABgene 1.2 mL Storage Plates, Black, are ideal for suspension cell culture and are compatible with automated systems. These plates come with either round or square shapes, and they are made of polypropylene. Different sealing options are available for the 1.2 mL Storage Plates, and you can seal each well independently to prevent cross-contamination.

Features of Thermo Scientific ABgene Storage Plate, 96-well, 1.2 mL, U-bottomed, Black

• Round well shape designed for optimal sample recovery
• Each well has an independent sealing rim to prevent cross-contamination
• Black color ideal for fluorescence-based assays
• Individually wrapped for convenience
• U-bottomed wells ideally suited for sample resuspension
• Polypropylene
• Resistant to DMSO (100%); Ethanol (100%); Isopropanol (100%)
• Suitable for Autoclaving (15 minutes at 121°C) or Gamma Irradiation
• Sealing options: Adhesive Seals, Heat Seals, Storage Plate Caps and Cap Strips, and Storage Plate Sealing Mats
• Cleanroom manufactured
• ANSI-format for compatibility with automated systems

Applications
• Sample storage
• Sample resuspension
• Cell culture
• Assay setup: diluting and aliquoting
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: 96-well plate
Plate Type: Storage Plate
Product Size: 50 plates
Shipping Condition: ELMS

Documents

Manuals & protocols