This product is only available through your local Thermo Scientific supplier. Learn where to buy >
Notice: This product will be discontinued on September 1, 2017. For superior performance, we recommend DreamTaq Green PCR Master Mix (2X).

ThermoPrime 2x ReddyMix PCR Master Mix, with 1.5 mM MgCl2

Catalog number: AB0575DCLDA

Thermo Scientific™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific ThermoPrime Taq DNA Polymerase with 10X ReddyMix PCR Buffer. This unique buffer has an inert red dye and a density reagent added. After thermal cycling, a sample of the PCR mix may be removed and loaded directly onto an agarose gel without the addition of loading dye. ReddyMix has no adverse effect on the outcome of PCR; yields are the same as with standard ThermoPrime Taq DNA Polymerase. Convenient Master Mix format allows easier reaction set-up.

Features

• Enhanced enzyme stability – for increased yield
• Saves time – no need to add gel loading dye
• Advanced buffering system – minimizes PCR optimization steps
• Convenient Master Mix formats – save time and reduce the risk of sample contamination during assay setup

Applications

• Amplifying fragments up to 5 kb
• Routine PCR

Related Products
ThermoPrime Taq DNA Polymerase, with 10x ReddyMix PCR buffer and separate vial of 25 mM MgCl2
ThermoPrime 2x ReddyMix PCR Master Mix, with 1.5 mM MgCl2
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: ThermoPrime Taq DNA Polymerase
Hot Start: No
Fidelity (vs. Taq): 1 X
GC-Rich PCR Performance: Low
Reaction Format: SuperMix or Master Mix
Product Overhang: 3'-A
Reaction Speed: Standard
Product Line: ThermoPrime
Product Size: 160 reactions

Contents & storage

Contains:
• 2 x 1 mL ReddyMix PCR Master Mix with 1.5 mM MgCl2 (2X)

Store at -20°C.

Documents

Manuals & protocols