Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils

Nunc™ Seals, Tapes, and Foils

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialFor Use With (Application)Certified RNase/DNase-free
AB0580PVC-coated PolypropyleneCrystallography and genotypingNo
232698AluminumPCR, storage in 96-well platesYes
232699AluminumPCR, storage in 384 well platesYes
232701PolypropylenePCRYes
232702PolypropylenePCR, fluorescence assays, microscopyYes
235306PolyethyleneLow autofluorescence, ELISANo
235307PolyolefinPCR, qPCR, ELISA, Protein crystallographyYes
236272VinylLuminescence, photosensitive samplesNo
236366PolyesterCell culture, sterile applicationsNo
236366PKPolyesterCell culture, sterile applicationsNo
236370PolyesterGeneral purpose, sample detectionNo
236703VinylFluorescence, photosensitive samplesNo
236707PolyesterGeneral purpose, ELISAYes
236710Polyolefin AcrylateLow autofluorescenceYes
241205Non-woven RayonBreathable, Cell cultureNo
249720Non-woven RayonCell growth, Bacterial growthYes
250050PlasticConsistent sealNo
276014AluminumPCR, long-term storageYes
3501PolyesterShort-term storage, ELISANo
5701TS1PolyesterShort-term storage, ELISA, minimizes evaporation and cross contaminationNo
AB0718PolypropyleneVentilating and storing cell culturesNo
Catalog number AB0580
Price (USD)
128.23
Online exclusive
149.65
Save 21.42
Case of 100
Material:
PVC-coated Polypropylene
For Use With (Application):
Crystallography and genotyping
Certified RNase/DNase-free:
No
Price (USD)
128.23
Online exclusive
149.65
Save 21.42
Case of 100

Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AutoclavableAutoclavable
DescriptionAdhesive Plate Seals
For Use With (Application)Crystallography and genotyping
Incubation ConditionsGeneral
No. per Case100
No. per Pack100
PeelableYes
Product TypeAdhesive Seal
Solvent Or Alcohol ResistantLife Science Reagent Resistant
Temperature Range (Metric)-40°C to +80°C
Well ShapeAll well shapes
AdhesiveAcrylic
Certified RNase/DNase-freeNo
ColorClear
MaterialPVC-coated Polypropylene
No. of Wells96
PierceableYes
Plate CompatibilityAll Microplates
Product LineAbgene
SterilityNon-sterile
Unit SizeCase of 100
Contents & Storage
General; For long or short term storage

Frequently asked questions (FAQs)

Are Thermo Scientific adhesive seals free of DNase, RNase, and human DNA?

Whilst Thermo Scientific adhesive seals are manufactured in a clean facility, they are not certified free of DNase, RNase, and human DNA.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

I am having problems with evaporation when using an adhesive seal. What do you recommend?

To prevent evaporation when using adhesive seals, firm application pressure must be applied to establish robust adhesive contact and bond strength around well rims. Adhesive seals such as ABsolute qPCR Seal (Cat. No. AB-1170) require sufficient pressure due to their non-tacky, pressure-sensitive adhesive. Adhesive Seal Applicator (Cat. No. AB-1391) or a similar tool may be used for optimal application of the seals. Alternatively, we recommend Thermo Scientific VersiCap Mats which show low sample evaporation with the performance of a cap. VersiCap Mats (Cat. Nos. AB-1810, AB-1815, AB-1820) offer comparable convenience and are easier to apply uniformly than adhesive seals.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.