PCR Plate, 96-well, non-skirted, white - FAQs
PCR Plate, 96-well, non-skirted, white - FAQs
View additional product information for PCR Plate, 96-well, non-skirted, white - FAQs (AB0600W)
1 product FAQs found
Why should I use Thermo Scientific white plastics in qPCR?
White plates and tubes reflect significantly more signal back to the qPCR instrument's detector than traditional clear plates. The improved signal reflection ensures that even the lowest levels of fluorescence are detected. White well walls also prevent signal from passing through to the cycler block where it can be inconsistently reflected or absorbed. This keeps variations in the cycler block from affecting qPCR data. Using Thermo Scientific white PCR plastics increases sensitivity and reduces variability within the qPCR assay.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.