Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Thermo-Fast PCR Plate, 24-well
Thermo Scientific™

Thermo-Fast PCR Plate, 24-well

Thermo Scientific 24-Well Semi-Skirted PCR Plates are pre-cut into 24-well segments. Their convenient format allows for lower sample numbers. TheRead more
Have Questions?
Catalog NumberQuantity
AB062450 plates
Catalog number AB0624
Price (USD)
154.65
Online exclusive
227.00
Save 72.35
Each
Quantity:
50 plates
Request bulk or custom format
Price (USD)
154.65
Online exclusive
227.00
Save 72.35
Each
Thermo Scientific 24-Well Semi-Skirted PCR Plates are pre-cut into 24-well segments. Their convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding.

Features of Thermo Scientific 24-Well Semi-Skirted PCR Plates

• Conveniently pre-cut into 24-well segments
• Suitable for use in 0.2 mL thermal cycler blocks
• Semi-skirt adds rigidity and allows for labeling or barcoding
• 0.3 mL maximum well volume (when used with adhesive seals)
• Sealable using adhesive films and foils and all cap strips

Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.

Related Products
Thermo-Fast PCR Plate, 24-well, green
Thermo-Fast PCR Plate, 24-well, purple
Thermo-Fast PCR Plate, 24-well, red
Thermo-Fast PCR Plate, 24-well, yellow
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Thermal Cycler
Packaging50 Plate
Product TypePCR Microplate
Quantity50 plates
Reaction SpeedStandard
Skirt StyleSemi
Volume (Metric) Working0.2 mL
BarcodeNo Barcode
ColorClear
MaterialPolypropylene
No. of Wells24
Unit SizeEach

Frequently asked questions (FAQs)

Plate warping is causing problems with the robot in my automated processes. How can I solve this?

We recommend Thermo Scientific Armadillo PCR plates which are designed for robotic handling with high rigidity and resistance. Combining the rigidity of a polycarbonate frame with thin walled polypropylene wells, the Armadillo plates provide superior thermal cycling performance under all conditions without warping.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics endotoxin-free?

They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics sterile?

Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What 96-well plates do you recommend for -80 degrees C storage?

Both polypropylene and polystyrene plates can be used to store samples at -80 degrees C and depends on the downstream application. Please note that the sealing method selected is critical for storing at these temperatures and heat sealing is recommended.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Which caps are Thermo-Fast PCR Plate, 24-well (Cat. No. AB0624) compatible with?

This plate is compatible with all caps e.g. Flat PCR Caps, strips of 8 (Cat. No. AB0784) and Domed PCR Caps, strips of 8 (Cat. No. AB0265).

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.