Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates

Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates

Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and pierceable formats.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)Well Shape
AB0674-20°C to +130°CRound
AB0566-20°C to +55°CRound
AB0675
also known as AB-0675
-20°C to +130°CSquare
AB0566SP-20°C to +120°CRound
Catalog number AB0674
Price (USD)
181.18
Online exclusive
253.75
Save 72.57
Case of 50
Temperature Range (Metric):
-20°C to +130°C
Well Shape:
Round
Price (USD)
181.18
Online exclusive
253.75
Save 72.57
Case of 50
Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and pierceable formats.

Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either pierceable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.

Highlights

  • Certified RNAse, DNAse, PCR Inhibitor, and human DNA free
  • Round or square capped mats for sealing 96-well storage plates
  • Alphanumeric grid-referencing
  • Molded from a proprietary thermoplastic elastomer for a flexible design
  • Suitable for long term storage
  • Available in autoclavable or pierceable formats

Pierceable version:

  • Solvent resistant (including DMSO, Ethanol, and Isopropanol)
  • Suitable for Gamma Irradiation
  • Requires 6N for piercing
  • Temperature Range: -20°C to +55°C

Autoclavable version:

  • Suitable for Gamma Irradiation
  • Temperature Range: -20°C to +130°C

Hazardous: No

Storage Condition: Ambient

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
DescriptionCapMat for 96-well Polypropylene Sample Processing & Storage Plates
For Use With (Application)Designed for sealing 96-well storage plate and suitable for long term storage
Format96-well
Green FeaturesACT Ecolabel (Silver)
No. per Case50
No. per Pack10
Plug Diameter7 mm
Product Type96-well capmat, sealing mat
Solvent Or Alcohol ResistantAlcohol Resistant, Solvent Resistant
Temperature Range (English)-4°F to 266°F
Temperature Range (Metric)-20°C to +130°C
Well ShapeRound
AdhesiveNone
Certified RNase/DNase-freeYes
ColorNatural
Fit for PurposePCR Ready
MaterialTPE
No. of Wells96
PierceableNo
Plate CompatibilityAbgene 96 Round Well Plate
Product LineAbgene
SterilitySterile
Unit SizeCase of 50