Abgene™ Plate Covers
Abgene™ Plate Covers
Abgene™ Plate Covers
Abgene™ Plate Covers

Abgene™ Plate Covers

Polypropylene 96-well plate covers, with or without ridges for easy stacking
Have Questions?
Catalog NumberPlate Compatibility
AB0755
also known as AB-0755
Abgene 96 and 384-well Plate
Catalog number AB0755
also known as AB-0755
Price (USD)
94.09
Online exclusive
109.62
Save 15.53
Case of 50
Plate Compatibility:
Abgene 96 and 384-well Plate
Price (USD)
94.09
Online exclusive
109.62
Save 15.53
Case of 50
Polypropylene 96-well plate covers, with or without ridges for easy stacking

Thermo Scientific Polypropylene Plate Covers with stacking ridges provide effective sealing options for a variety of applications and can be used with 96-well plates.

Highlights

  • Ridged covers for stacking of covered plates
  • Non-ridged covers for use with automated systems
  • Polypropylene covers, ridged and non-ridged, are available for use with and 384-well plates

Hazardous: No

Storage Condition: Ambient

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Condensation RingsNo
Cut-off CornersH1 Corner
DescriptionMicroplate Lid
Dimensions (L x W)5 x 3.31 in. (126 x 84 mm)
Dimensions (L x W x H)5 x 3.31 x 0.36 in. (126 x 84 x 9.25mm)
FormatLid
IncludesWith stacking ridge
No. per Case50
No. per Pack2
Product TypeMicroplate Lid, cover
Temperature Range (Metric)-20°C to +120°C
Well ShapeAll well shapes
AdhesiveNone
ColorNatural
Fit for PurposePCR Ready
MaterialPolypropylene
No. of Wells96
Plate CompatibilityAbgene 96 and 384-well Plate
Product LineAbgene
SterilityCleanroom
Unit SizeCase of 50