How important is the quality of my DNA template in long PCR?
Template preparation becomes particularly important when performing longer amplifications (>15 kb). Therefore, it is recommended to check the length of the DNA by agarose gel electrophoresis.