Notice: This product will be discontinued on May 1, 2017. For superior performance, we recommend Phusion Green Hot Start II High-Fidelity PCR Master Mix.

Extensor Long PCR ReddyMix Master Mix, with Buffer 1

Catalog number: AB0794B

Thermo Scientific™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Extensor Long Range PCR Enzyme is a blend of ThermoPrime Taq DNA Polymerase and a proprietary thermostable proofreading enzyme. The two enzymes act synergistically to generate long PCR products, up to 20 kb. This enzyme blend is available in 2X Master Mix and ReddyMix formulations. The ReddyMix Master Mix contains an inert red dye and a density reagent that allows for direct loading onto agarose gels.

Highlights

• Proprietary enzyme blend amplifies targets up to 20 kb in length
Applications

• Long range PCR
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: Extensor Long Range PCR Enzyme
Hot Start: No
Fidelity (vs. Taq): 6 X
GC-Rich PCR Performance: Medium
Reaction Format: SuperMix or Master Mix
Product Overhang: Mixed
Reaction Speed: Standard
Product Line: Extensor
Product Size: 400 reactions

Contents & storage

Contains:
• 5 x 1 mL Extensor PCR ReddyMix Master Mix with Buffer 1 (2X), which includes
— Extensor Long Range PCR Enzyme,
— Buffer 1,
— dNTPs, and
— MgCl2 in ReaddyMix format.

Store at -20°C.

Documents

Manuals & protocols