Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Thermo Scientific™

Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates

384-well microarray plate with pyramidal well bottoms for maximum sample recovery.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorVolume (Metric) WellWell ShapeFit for Purpose
AB0862Natural300 μLSquare, Conical Bottom (V-well)PCR Ready
AB0781Natural120 μLSquare, Conical Bottom (V-well)PCR Ready
AB1178Natural250 μLSquare, Conical Bottom (V-well)PCR Ready
AB1056
also known as AB-1056
Natural55 μLRound, Conical Bottom (V-well)PCR Ready
Catalog number AB0862
Price (USD)
807.00
Case of 50
Color:
Natural
Volume (Metric) Well:
300 μL
Well Shape:
Square, Conical Bottom (V-well)
Fit for Purpose:
PCR Ready
Price (USD)
807.00
Case of 50

Thermo Scientific™ Abgene™ polypropylene microplates and DeepWell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use and have molded slots and pads for automated sample processing and preparation workflows. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using USP <661.1> high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and DeepWell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.

All Thermo Scientific plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.

 

Highlights

  • Certified free from DNAse, RNAse, PCR Inhibitors, and human DNA
  • Pyramidal well bottom for ease of sample retrieval
  • Discrete raised rims around each well for effective sealing and to reduce the risk of cross-contamination

Compatible Sealing Film: Adhesive and Heat
Hazardous: No
Max Centrifugal Force: 2000-5000 RCF
Storage Condition: Ambient stored, or transported

All plates are ANSI format for compatibility with automated systems.

To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.

Specifications
AutoclavableAutoclavable
Binding TypeLow binding PP
Certifications/ComplianceCertified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Dnase FreeYes
Description384-well Polypropylene Sample Processing & Storage Plates
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
FormatStandard
Max. RCF5000 x g
No. per Case50
No. per Pack50
Packaging48 Pk.
Plate SizeANSI/ SLAS Standard
Product Type384-well plate
RNase FreeYes
Regulatory StatusRUO
Volume (Metric) Well300 μL
Volume (Metric) Working250 μL
Array16 x 24
BarcodeCustom Barcoding Available
ColorNatural
Fit for PurposePCR Ready
MaterialPolypropylene
No. of Wells384
Product LineAbgene
SterilityCleanroom
Surface TreatmentNon-treated
Well ShapeSquare, Conical Bottom (V-well)
Unit SizeCase of 50

Frequently asked questions (FAQs)

Are Thermo Scientific ABgene plastics endotoxin-free?

They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics sterile?

Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What 96-well plates do you recommend for -80 degrees C storage?

Both polypropylene and polystyrene plates can be used to store samples at -80 degrees C and depends on the downstream application. Please note that the sealing method selected is critical for storing at these temperatures and heat sealing is recommended.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What is the recommended storage temperature for Abgene 384 Well Polypropylene Sample Processing & Storage Plates?

We recommend storing Abgene 384 Well Polypropylene Sample Processing & Storage Plates at ambient temperature.