Ultra Clear qPCR Caps, strips of 8 - FAQs
Ultra Clear qPCR Caps, strips of 8 - FAQs
View additional product information for Ultra Clear qPCR Caps, strips of 8 - FAQs (AB0866)
2 product FAQs found
I would like to use the flat ultra-clear and domed cap strips for Thermo-Fast PCR Plate, 48-well, clear (Cat. No. AB0648). What do you recommend?
For the domed cap strips, you could choose the Domed PCR Caps, strips of 8 (Cat. No. AB0265). For the flat ultra clear cap strips you can choose for the flat ultra clear cap strips the (Cat. No. AB0866). Generally speaking, within the Thermo Scientific/ABgene legacy, all our cap strips are compatible with all PCR plates and tube strips. For more options, please look at the following link.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
Can I order the Caps within the Low Profile Tubes (white) and Ultra Clear Caps, strips of 8 (Cat. No. AB1771) seperately?
Yes. The suitable stand alone products are either the Ultra Clear qPCR Caps, strips of 8 (Cat. No.AB0866) or the Domed PCR Caps, strips of 8 (Cat. No. AB0265).