Abgene Storage Plate Caps, Strips of 8
Thermo Scientific™

Abgene Storage Plate Caps, Strips of 8

Storage Plate Caps and Mats are available in a variety of formats.
Have Questions?
Catalog NumberColor
AB0981
also known as AB-0981
Natural
Catalog number AB0981
also known as AB-0981
Price (USD)
64.95
Online exclusive
75.82
Save 10.87
Case of 120
Color:
Natural
Price (USD)
64.95
Online exclusive
75.82
Save 10.87
Case of 120
Storage Plate Caps and Mats are available in a variety of formats.

Thermo Scientific storage plate cap strips are suitable for use on with Round Well Storage Plates and feature a low-profile design. They come in an 8-cap strip and can simultaneously seal up to 8 well. Each cap is individually numbered.

Highlights

  • 8-cap strip
  • Each cap is individually numbered
  • Simultaneous sealing of 8 wells
  • End tabs for ease of removal
  • Low profile for increased well volume

Autoclavable- Resistance: Not Recommended

DMSO (100%) - Resistance: Successfully Tested

Ethanol (100%) - Resistance: Successfully Tested

Gamma Irradiation - Resistance: Successfully Tested

Hazardous: No

Isopropanol (100%) - Resistance: Successfully Tested

Long Term Strorage: Successfully Tested

Peelable: Successfully Tested

Pierceable:Not Recommended

Resealable: Successfully Tested

Sealing Temp Range: -20°C to +55°C

Storage Condition: Ambient

UV Irradiation- Resistance: Successfully Tested

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AutoclavableAutoclavable
Product TypePlate Lid
Well ShapeRound
Certified RNase/DNase-freeYes
ColorNatural
MaterialPolypropylene
No. of Wells8
PierceableNo
Plate CompatibilityAbgene 96 Round Well Plate
Product LineAbgene
SterilitySterile
Unit SizeCase of 120