What is the difference between PCR Plate, 96-well, low profile, skirted (Cat. No. AB0800, AB0800150) and PCR Plate, 96-well, robotic (Cat. No. AB1300)?
There is a slight difference in the height of the PCR plates. PCR Plate, 96-well, low profile, skirted (Cat. No. AB0800, AB0800150) is 15.5 mm tall and PCR Plate, 96-well, robotic (Cat. No. AB1300) is 17.0 mm tall.