PCR Plate, 384-well, standard, purple
Thermo Scientific™
PCR Plate, 384-well, standard, purple
The Standard Thermo Scientific™ 384-well plate, purple, is compatible with the majority of 384 block PCR and sequencing instruments.
Have Questions?
Número de catálogoCantidad
AB1384P50 placas
Número de catálogo AB1384P
Precio (MXN)
-
Cantidad:
50 placas
Pedido a granel o personalizado
The Standard Thermo Scientific™ 384-well plate, purple, is compatible with the majority of 384 block PCR and sequencing instruments. It has a maximum well volume of 40 μL and a working volume of 25 μL. This 384-well plate is compatible with all major 384 block thermal cyclers, and it has a cut corner at the A24 location.
Highlights of the Standard Thermo Scientific™ 384-well plate
  • Fully skirted for use with automated systems
  • Purple color to aid in sample tracking
  • Compatible with all leading 384 block thermal cyclers
  • 40 μL maximum well volume, 25 μL working volume
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
  • Cut corner: A24
Applications
  • PCR
  • qPCR/Real-Time PCR
Especificaciones
Certificaciones/conformidadCertificación como material libre de ARNasa y ADNasa
Para utilizar con (equipo)Sistemas de PCR en tiempo real o termocicladores
Tipo de productoMicroplaca de PCR
Cantidad50 placas
Velocidad de reacciónEstándar
Con rebordeSí
Estilo de faldaCon faldón
Volumen del pocillo (métrico)40 μl
Volumen de trabajo (métrico)25 μl
Código de barrasSin código de barras
ColorMorado
MaterialPolipropileno
N.° de pocillos384
Unit SizeEach