PCR Plate, 384-well, standard, red
Thermo Scientific™

PCR Plate, 384-well, standard, red

The Standard Thermo Scientific 384-well plate, red, is compatible with the majority of 384 block PCR and sequencing instruments. ItRead more
Have Questions?
Catalog NumberQuantity
AB1384R50 plates
Catalog number AB1384R
Price (USD)
442.00
Each
Quantity:
50 plates
Price (USD)
442.00
Each
The Standard Thermo Scientific 384-well plate, red, is compatible with the majority of 384 block PCR and sequencing instruments. It has a maximum well volume of 40 μL and a working volume of 25 μL. This 384-well plate is compatible with all major 384 block thermal cyclers, and it has a cut corner at the A24 location.

Highlights of the Standard Thermo Scientific 384-well plate

• Fully skirted for use with automated systems
• Red color to aid in sample tracking
• Compatible with all leading 384 block thermal cyclers
• 40 μL maximum well volume, 25 μL working volume
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer
• White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Cut corner: A24

Applications

• PCR
• qPCR/Real-Time PCR
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging50 Plate
Product TypePCR Microplate
Quantity50 plates
Reaction SpeedStandard
Skirt StyleSkirted
Volume (Metric) Working25 μL
BarcodeNo Barcode
ColorRed
MaterialPolypropylene
No. of Wells384
Unit SizeEach