PCR Plate, 384-well, standard, yellow
Thermo Scientific™

PCR Plate, 384-well, standard, yellow

The Standard Thermo Scientific™ 384-well plate, yellow, is compatible with the majority of 384 block PCR and sequencing instruments.
Have Questions?
Catalog NumberQuantity
AB1384Y50 Plates
Catalog number AB1384Y
Price (MXN)
-
Quantity:
50 Plates
Request bulk or custom format

The Standard Thermo Scientific™ 384-well plate, yellow, is compatible with the majority of 384 block PCR and sequencing instruments. It has a maximum well volume of 40 μL and a working volume of 25 μL. This 384-well plate is compatible with all major 384 block thermal cyclers, and it has a cut corner at the A24 location.

Highlights of the Standard Thermo Scientific™ 384-well plate

  • Fully skirted for use with automated systems
  • Yellow color to aid in sample tracking
  • Compatible with all leading 384 block thermal cyclers
  • 40 μL maximum well volume, 25 μL working volume
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
  • Cut corner: A24

Applications

  • PCR
  • qPCR/Real-Time PCR
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Product TypePCR Microplate
Quantity50 Plates
Reaction SpeedStandard
RimmedYes
Skirt StyleSkirted
Volume (Metric) Well40 μL
Volume (Metric) Working25 μL
BarcodeNo Barcode
ColorYellow
MaterialPolypropylene
No. of Wells384
Unit SizeEach