What are the benefits of using EasyStrip Plus Tube Strip with Attached Flat Caps?
These new strips have new features including a graduated 20 µL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.