Armadillo PCR Plate, 384-well, blue, clear wells - FAQs
Armadillo PCR Plate, 384-well, blue, clear wells - FAQs
View additional product information for Armadillo PCR Plate, 384-well, blue, clear wells - FAQs (AB2384B)
1 product FAQs found
Are the Armadillo PCR Plate, 384-well, blue, clear wells (Cat. No. AB2384B) compatible with the QuantStudio 6 Pro having a 384 well block?
Yes, they are compatible. For checking the compatibility of the plastics to your instrument, please see the PCR and qPCR Plastics Selection Tool.