Do I need to treat my RNA with DNase I before running RT-qPCR with Verso cDNA Synthesis Kit?
No, you do not need to treat your RNA with DNase I before running RT-qPCR with the Verso cDNA Synthesis Kit. The RT enhancer included in the Verso kit degrades any contaminating double-stranded DNA during the reverse transcription process, eliminating the need for a separate DNase I treatment.
Can I purchase the Verso Enzyme Mix separately from the Verso 1-step RT-qPCR Kit (Cat. No. AB4016A, AB4016C)?
No, we do not offer the Verso Enzyme Mix as a standalone product. It is only available as a component of the Verso 1-step RT-qPCR Kit (Cat. No. AB4016A, AB4016C).