Orion™ AQUAfast™ II Chemistries Test Kits
Thermo Scientific™

Orion™ AQUAfast™ II Chemistries Test Kits

Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeRangeQuantity
AC2065Zinc0.02 to 1 mg/L Zn50 Tests
AC2002Alkalinity5 to 200 mg/L CaCO3100 Tests
AC3002PCaCO35 to 300 mg/L100 Tests
AC2027Aluminum0.01 to 0.3 mg/L Al50 Tests
AC4P27Aluminum0.01 to 0.25 mg/L100 Tests
AC2012Ammonia0.02 to 1 mg/L N50 Tests
AC2035Bromine Reagent Tablet0.1 to 13.5 mg/L
AC2017Chloride0.5 to 25 mg/L Cl-50 Tests
AC2099Chlorine Dioxide0.02 to 11 mg/L ClO2100 Tests
AC2071Chlorine0.01 to 6 mg/L Cl2100 DPD No 1 Tablets
AC2070Chlorine0.01 to 6 mg/L Cl250 Tests Each (free and total)
AC2072Chlorine0.01 to 6 mg/L Cl2100 DPD No 4 Tablets
AC3072Chlorine, Sodium Chlorate5 to 200 mg/L100 Tests
AC2029Copper0.05 to 5 mg/L Cu50 Tests
AC2098Cyanuric Acid5 to 90 mg/L100 Tests
AC3032C2Calcium0 to 500 mg/L100 Tests
AC3032CCalcium50 to 900 mg/L100 Tests
AC3032TMetallphthalein Method2 to 50 mg/L100 Tests
AC2030Hydrazine0.05 to 0.5 mg/L N2H430 Tests
AC3069Hydrogen Peroxide0.03 to 3 mg/L100 Tests
AC2078Iron0.02 to 1 mg/L Fe100 Tests
AC2055Manganese0.2 to 4 mg/L Mn50 Tests
AC2007Nitrate1 to 40 mg/L50 Tests
AC2046Nitrite0.01 to 0.5 mg/L N100 Tests
AC2048Ozone0.05 to 0.5 mg/L O3100 Tests
AC2001pH6.5 to 8.4 pH100 Tests
AC3001BPBromocresol Purple, pH, Low Range5.2 to 6.8 pH100 Tests
AC3001TBThymol Blue, pH, High Range8.0 to 9.6 pH100 Tests
AC2095-WAPhosphate0.05 to 4 mg/L PO450 Tests
AC2096Phosphate1 to 80 mg/L PO450 Tests
AC3019Potassium0 to 12 mg/L100 Tests
AC2060Silica0.05 to 4 mg/L SiO250 Tests
AC2061Silica0.05 to 4 mg/L SiO2100 Tablets
AC2082Sulfate5 to 100 mg/L SO4100 Tests
AC2016Sulfide0.04 to 0.5 mg/L S100 Tests
Catalog number AC2065
Price (USD)
46.89
Online exclusive
52.10
Save 5.21
Each
Type:
Zinc
Range:
0.02 to 1 mg/L Zn
Quantity:
50 Tests
Price (USD)
46.89
Online exclusive
52.10
Save 5.21
Each

Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
  • No. of tests: 50 to 100

Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionAQUAfast zinc reagent tablets, 50 tests
EPA Approved MethodsWastewater: Standard Methods 3500-Zn B
For Use With (Application)Colorimetric water and wastewater testing equipment
IncludesSet of 2: 50 Copper/Zinc LR Tablets, 50 EDTA Table
MethodZincon
Quantity50 Tests
Range0.02 to 1 mg/L Zn
Recommended AccessoriesAQUAfast 24mm round vials (Cat. No. AC2V24)
Test Reagent FormatsTablet
For Use With (Equipment)Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
TypeZinc
Unit SizeEach