Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Actual product may vary
Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Thermo Scientific™

Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents

Perform colorimetric testing including fluoride, COD, total nitrogen and total phosphate using the Thermo Scientific™ Orion™ AQUAfast Liquid Reagents, Digestion and Reaction Tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeRange
AC3001pH6.5 to 8.4 pH
AC2009Fluoride0.05 to 2 mg/L F
AC4P54Manganese0.01 to 0.7 mg/L Mn
ACD004Nitrogen, Total0.5 to 25 mg/L N
ACD007Nitrogen, Total5 to 150 mg/L N
ACD095Phosphate0.02 to 1.1 mg/L P
ACD095AHPhosphate0.02 to 1.6 mg/L P
ACR007Nitrate1 to 30 mg/L N
ACR011Ammonia1 to 50 mg/L N
ACR012Ammonia0.02 to 2.5 mg/L N
ACR095Phosphate0.06 to 5 mg/L PO4
CODH00COD0 to 1500 mg/L O2
CODHP0COD0 to 15000 mg/L O2
CODL00COD0 to 150 mg/L O2
CODH150COD High Range0 to 15000 mg/L O2
CODL150COD Low Range0 to 150 mg/L O2
CODM150COD Mid Range0 to 1500 mg/L O2
Catalog number AC3001
Price (USD)
51.66
Online exclusive
57.40
Save 5.74
Each
Type:
pH
Range:
6.5 to 8.4 pH
Price (USD)
51.66
Online exclusive
57.40
Save 5.74
Each
Use Thermo Scientific™ Orion™ AQUAfast Digestion Tubes, Reaction Tubes and Liquid Reagents with Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. Digestion and reaction tubes contain the precise amount of reagent, are made of special optical glass and fit the Orion™ AQUAfast COD165 thermoreactor. Digestion tubes measure COD, total nitrogen and total phosphate. Reaction tubes measure ammonia, nitrate and phosphate. Liquid reagents mix quickly and easily in solution and measure fluoride (SPADN), hydrazine and pH.
Changes in nutrient levels can alter the delicate balance in ecosystems and contaminate drinking water. Tube tests make it easy to determine concentrations of species in samples.

 

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Digestion tube tests and reaction tube tests are preprogrammed in the Orion AQ3700 meter. Some of the listed tests have only liquid components, like our AC2009 SPADNS test for fluoride, and some have liquid and powder or liquid and tablet reagents.
Specifications
DescriptionAQUAfast pH liquid dropper reagent
For Use With (Equipment)Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ3700 Colorimeter, Orion AQUAfast
FormatLiquid
IncludesPhenol Red Solution
Quantity30 Tests
Range6.5 to 8.4 pH
Recommended AccessoriesAQUAfast 24mm round vials (Cat. No. AC2V24)
Test MethodPhenol Red
TypepH
Unit SizeEach