Product Image
Thermo Scientific™
Turbidity Standards for Orion™ AQ3010 Turbidity Meter
Replacement Turbidity Standards for the Orion AQ3010 Turbidity meter.
Have Questions?
Catalog NumberFor Use With (Equipment)
AC301SOrion AQ3010 Turbidity Meter
Catalog number AC301S
Price (USD)
314.10
Online exclusive
349.00
Save 34.90
Each
For Use With (Equipment):
Orion AQ3010 Turbidity Meter
Price (USD)
314.10
Online exclusive
349.00
Save 34.90
Each
Turbidity Standards are for use with the Orion™ AQ3010 Turbidity Meter (Cat. No. AQ3010). These replacement calibration standards kit are polymer-based primary turbidity standards and includes 800, 100, 20 and 0.02 NTU in 60mL plastic bottles.
  • For use with: Orion AQUAfast AQ3010
  • Standards Set has 60 mL bottle each of 0.02, 20.0, 100, and 800 NTU 100
Specifications
DescriptionReplacement calibration standards kit, includes 800, 100, 20.0 and 0.02 NTU standards in plastic bottles
Includes800, 100, 20.0 and 0.02 NTU standards in 60mL bottles
For Use With (Equipment)Orion AQ3010 Turbidity Meter
TypeCalibration Standards
Unit SizeEach