Orion™ Powder Chemistry Packs
Orion™ Powder Chemistry Packs
Orion™ Powder Chemistry Packs
Thermo Scientific™

Orion™ Powder Chemistry Packs

Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeRange
AC4P95Phosphate (ortho)0.06 to 2.5 mg/L PO4
AC4P12Ammonia0.01 to 0.8 mg/L N
AC4P29Copper (free)0.05 to 5 mg/L Cu
AC4P42Molybdate/Molybdenum0.5 to 66 mg/L MoO4
AC4P46Nitrite0.01 to 0.3 mg/L N
AC4P55Manganese0.1 to 18 mg/L Mn
AC4P60Silica1 to 90 mg/L SiO2
AC4P71Chlorine (free)0.02 to 2 mg/L Cl2
AC4P72Chlorine (total)0.02 to 2 mg/L Cl2
AC4P78Iron (ferro)0.02 to 3 mg/L Fe
AC4P79Iron (total)0.02 to 1.8 mg/L Fe
AC4P82Sulfate5 to 100 mg/L SO4
Catalog number AC4P95
Price (USD)
51.66
Online exclusive
57.40
Save 5.74
Each
Type:
Phosphate (ortho)
Range:
0.06 to 2.5 mg/L PO4
Price (USD)
51.66
Online exclusive
57.40
Save 5.74
Each

Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.

  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast™ Colorimeters

Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
EPA Approved MethodsDrinking Water: ASTM D515-88 (A) / Standard Methods 4500-P E; Wastewater: Standard Methods 4500-P E
For Use With (Equipment)Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter
Includes100 Phos3 Powder Packs
PackagingPowder Pack
Quantity100 tests
Range0.06 to 2.5 mg/L PO4
Recommended AccessoriesAQUAfast 24 mm round vials (Cat. No. AC2V24)
Test MethodPhosphomolybdenum/Ascorbic Acid
TypePhosphate (ortho)
Unit SizeEach