This product is no longer available to order. Alternative product Food Safety Testing QuantStudio™ 5 Real-Time PCR System - A36328 - is available using the Thermo Scientific™ SureTect™ method AOAC certified and/or AFNOR validated workflow. Click the link to find out more.
Pathatrix™ Auto Cartridge Rack
Applied Biosystems™

Pathatrix™ Auto Cartridge Rack

Have Questions?
Catalog NumberQuantity
ACARTRACK1 each
Catalog number ACARTRACK
Price (USD)
430.00
Each
Quantity:
1 each
Price (USD)
430.00
Each
The Pathatrix™ Auto Cartridge Rack is designed for use with the Pathatrix™ Auto Instrument. The rack holds a set of 5 Cartridges and provides a steady base that simplifies loading and unloading of the plastic consumable kits into and out of the Cartridges.
Specifications
CapacityUp to 5 Pathatrix™ Auto Cartridges
For Use With (Equipment)Pathatrix™ Auto System
Product TypeAuto Cartridge Rack
Quantity1 each
Shipping ConditionRoom Temperature
Product LinePathatrix
Unit SizeEach
Contents & Storage
1 rack

Store at room temperature.