IL-2 Monoclonal Antibody (297C16F11) - FAQs
IL-2 Monoclonal Antibody (297C16F11) - FAQs
View additional product information for IL-2 Monoclonal Antibody (297C16F11) - FAQs (AHC0722)
1 product FAQs found
What is the neutralizing mechanism of Thermo Fisher Scientific' mouse anti-human IL-2 neutralizing antibody (Cat. No. AHC0722, clone 297C16F11)
Mouse monoclonal anti-IL-2 antibody, AHC0722, neutralizes cytokine activity by blocking an epitope on the IL-2 protein that normally binds to the p55 IL-2 receptor. This neutralizes the activity of the IL-2 protein.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.