Single Place Magnetic Stand (for one 1.5 mL tube) - FAQs
Single Place Magnetic Stand (for one 1.5 mL tube) - FAQs
View additional product information for Single Place Magnetic Stand (for one 1.5 mL tube) - FAQs (AM10026)
1 product FAQs found
Will Ambion magnetic stands (AM10050 & AM10026) work for 0.2 mL individual PCR tubes or strips?
Ambion magnetic stands (AM10050 & AM10026) are not designed for and do not work well for 0.2 mL individual PCR tubes or strips.