Magnetic-Ring Stand (96 well) - FAQs
Magnetic-Ring Stand (96 well) - FAQs
View additional product information for Magnetic-Ring Stand (96 well) - FAQs (AM10050)
2 product FAQs found
What magnetic stand should I purchase for manual extraction? Cat. No. AM10027 or Cat. No. AM10050?
It is a personal preference as to which magnetic stand to purchase. Cat. No. AM10027 can be easier if using a pipettor for liquid handling, as beads clump to the side, Cat. No. AM10050 if using a robotic liquid handler, as beads clump in a ring.
Will Ambion magnetic stands (AM10050 & AM10026) work for 0.2 mL individual PCR tubes or strips?
Ambion magnetic stands (AM10050 & AM10026) are not designed for and do not work well for 0.2 mL individual PCR tubes or strips.