RNAqueous™ Total RNA Isolation Kit
Invitrogen™
RNAqueous™ Total RNA Isolation Kit
The RNAqueous Total RNA Isolation Kit is for phenol-free total RNA isolation from 1–75 mg of tissue or from 10^2–10^7 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
AM10051G50 CatridgesSpin Columns
AM191250 PrepsRNA Isolation Kit
Catalog number AM10051G
Price (USD)
188.65
Online exclusive
196.00
Save 7.35
Each
Quantity:
50 Catridges
Product Type:
Spin Columns
Price (USD)
188.65
Online exclusive
196.00
Save 7.35
Each
The RNAqueous Total RNA Isolation Kit is for phenol-free total RNA isolation from 1–75 mg of tissue or from 102–107 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.
The RNAqueous kit provides:
  • Convenient syringe-based filtration
  • Compatibility with vacuum manifolds when processing multiple samples
How it works
The RNAqueous kit uses a powerful chaotropic solution for cell lysis, followed by RNA binding to glass fiber housed in a luer-lock syringe filter. Washing and elution solutions are simply pushed through the filter using a standard syringe (not included). Vacuum filtration is not required; however, the filter units are compatible with standard vacuum manifolds for processing several samples at once.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Product TypeSpin Columns
Quantity50 Catridges
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
50 cartridges; store at room temperature.
Frequently asked questions (FAQs)
Can the Filter Cartridges within the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908) be ordered separately?
Yes, we do offer the Spin Cartridges (Cat. No. AM10051G) as a stand-alone product, and they would be compatible with the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908).