Do you offer an alternative for the discontinued NucAway Spin Columns?
Yes, we recommend Spin Columns and Tubes (Cat. No. AM10065) (https://www.thermofisher.com/order/catalog/product/AM10065).
Search
View additional product information for Spin Columns and Tubes - FAQs (AM10065)
2 product FAQs found