Customer information regarding this product here »
Barrier (Filter) Tips, 100 μL size
Barrier (Filter) Tips, 100 μL size
Barrier (Filter) Tips, 100 μL size
Invitrogen™

Barrier (Filter) Tips, 100 μL size

Barrier tips prevent cross-contamination of samples by means of an aerosol filter. They are RNase- and DNase-free and are idealRead more
Have Questions?
Catalog NumberQuantity
AM1264810 racks
Catalog number AM12648
Price (USD)
259.35
Special offer
Online exclusive
Ends: 27-Jun-2026
273.00
Save 13.65 (5%)
10 racks
Quantity:
10 racks
Price (USD)
259.35
Special offer
Online exclusive
Ends: 27-Jun-2026
273.00
Save 13.65 (5%)
10 racks
Barrier tips prevent cross-contamination of samples by means of an aerosol filter. They are RNase- and DNase-free and are ideal for handling RNA, PCR applications, and for working with radioisotopes or toxic chemicals. Ten racks containing 8 x 12 tips each are provided. They are compatible with Eppendorf®, Finnpipette, Oxford Benchmate®, Pipetman™, Proline®, Rainin, Socorex, and other brands of pipettors. Each lot of tips is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 racks
Shipping ConditionRoom Temperature
AutoclavableAutoclavable
FilteredFiltered
For Use With (Equipment)Universal Fit
FormatRack
Product LineInvitrogen
SterilitySterile
Tip StyleUniversal Fit
Volume (Metric)100 μL
Unit Size10 racks
Contents & Storage
Store at room temperature.