Customer information regarding this product here »
Barrier (Filter) Tips, 200 μL size
Barrier (Filter) Tips, 200 μL size
Barrier (Filter) Tips, 200 μL size
Invitrogen™

Barrier (Filter) Tips, 200 μL size

Ambion® Barrier tips prevent cross-contamination of samples by means of an aerosol filter. They are RNase- and DNase-free and areRead more
Have Questions?
Catalog NumberQuantity
AM1265510 racks
Catalog number AM12655
Price (USD)
274.00
10 racks
Quantity:
10 racks
Price (USD)
274.00
10 racks
Ambion® Barrier tips prevent cross-contamination of samples by means of an aerosol filter. They are RNase- and DNase-free and are ideal for handling RNA, PCR applications, and for working with radioisotopes or toxic chemicals. Ten racks containing 8 x 12 tips each are provided. They are compatible with Eppendorf®, Finnpipette, Oxford Benchmate®, Pipetman™, Proline®, Rainin, Socorex, and other brands of pipettors. Each lot of tips is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 racks
Shipping ConditionRoom Temperature
AutoclavableAutoclavable
FilteredFiltered
For Use With (Equipment)Universal Fit
FormatRack
Product LineInvitrogen
SterilitySterile
Tip StyleUniversal Fit
Volume (Metric)200 μL
Unit Size10 racks
Contents & Storage
Store at room temperature.