The Ambion™ MAXIscript™ T7 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protectionRead more
Have Questions?
Change view
Catalog Number
Product Type
Quantity
AM1312
T7 In Vitro Transcription Kit
30 Reactions
AM1314
T7 In Vitro Transcription Kit
100 Reactions
AM1314M
T7 In Vitro Transcription Kit with Manual
100 Reactions
3 Options
Catalog number AM1312
Price (USD)
213.00
Each
Product Type:
T7 In Vitro Transcription Kit
Quantity:
30 Reactions
Price (USD)
213.00
Each
The Ambion™ MAXIscript™ T7 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations. Each kit contains sufficient reagents for 100 reactions.
Features:
• High yields of high specific activity, full-length RNA probes in just 10 minutes • Ideal probe synthesis kit for nuclease protection assays, northern and Southern blots, and in situ hybridization • Includes TURBO DNase™ • All kits contain quality-tested DNase/RNase-free reagents
The MAXIscript™ In Vitro Transcription Kit synthesizes RNA probes with specific activities reaching 1 × 109 cpm/μg in just 10 minutes. MAXIscript™ Kits are very efficient at synthesizing full-length probes, even at limiting nucleotide concentrations, making them the perfect companion to ribonuclease protection assays. MAXIscript™ Kits are also an excellent choice for the synthesis of nonisotopically labeled probes. Labeling can be accomplished post-transcriptionally by using Ambion's BrightStar™ Psoralen-Biotin Labeling Kit, or during transcription by incorporating modified nucleotides into the reaction. MAXIscript™ Kits can also be used for the synthesis of modest amounts of unlabeled RNA (2–6 μg/20 μl reaction) of up to 2 kb.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions30 Reactions
Product LineAmbion, MAXIscript
Product TypeT7 In Vitro Transcription Kit
PromoterT7
Quantity30 Reactions
Shipping ConditionDry Ice
Starting MaterialPCR Products, Linearized DNA plasmid
Yield2-6 μg
Final Product TypeSynthetic mRNA, mRNA
FormatKit
Sample TypemRNA, Synthetic mRNA
Unit SizeEach
Contents & Storage
T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Frequently asked questions (FAQs)
What probe kits do you offer for nonisotopic probe labeling and radiolabeled probe labeling?
We offer two nonisotopic probe labeling kits:
- The BrightStar Psoralen-Biotin Nonisotopic Labeling Kit (Cat. No. AM1480)
- MAXIscript Kits for in vitro transcription (Cat. Nos. AM1308 through AM1326)
We offer three radiolabeled probe labeling kits:
- MAXIscript Kits for in vitro transcription (Cat. Nos. AM1308 through AM1326)
- KinaseMax 5' End-Labeling Kit (Cat. No. AM1520)
- DECAprime II DNA Labeling Kit (Cat. No. AM1455)
What kits should I use to isolate RNA from my sample?
The best isolation kit to use will depend greatly on your application, but, in general, the kits below produce RNA input compatible with the Ion Total RNA-Seq Kit v2 workflow:
Whole Transcriptome (polyA-selected or rRNA-depleted RNA is highly recommended, but total RNA is acceptable)
- Poly A-selected RNA
Dynabeads mRNA DIRECT Micro Kit (Cat. No. 61021)
MicroPoly(A)Purist Kit (Cat. No. AM1919)
- rRNA-depleted RNA
RiboMinus Eukaryote System v2 (Cat. No. A15026)
Low Input RiboMinus Eukaryote System v2 (Cat. No. A15027)
- Total RNA
TRI Reagent
TRizol
RecoverAll MagMax