mMESSAGE mMACHINE® SP6 Transcription Kit with Manual - FAQs
mMESSAGE mMACHINE® SP6 Transcription Kit with Manual - FAQs
View additional product information for mMESSAGE mMACHINE® SP6 Transcription Kit with Manual - FAQs (AM1340M)
1 product FAQs found
What is the control included in the mMESSAGE mMACHINE kit?
The control is a pTRI-Xef control template which is a linearized TRIPLEscript plasmid contianing the 1.8 kb Xenopus elongation factor 1alpha gene under the transcription control of tandem SP6, T7, and T3 promoters (pTRI-Xef 1). Any one of the three polymerases can be used to synthesize the control RNA.