Do you offer kits for probe construction for small RNA and miRNA analysis?
Yes, we offer our mirVana miRNA Probe Construction Kit (Cat. No. AM1550) and our mirVana Probe and Marker Kit (Cat. No. AM1554).
Search
View additional product information for mirVana™ Probe & Marker Kit - FAQs (AM1554)
1 product FAQs found