I'd like to isolate RNA and protein from the same sample. Do you have a kit for this?
Yes, you can use TRIzol Reagent or the mirVana PARIS RNA and Native Protein Purification Kit.
Search
View additional product information for mirVana™ PARIS™ Kit with manual - FAQs (AM1556M)
1 product FAQs found