MessageAmp™ II-Bacteria RNA Amplification Kit - FAQs
MessageAmp™ II-Bacteria RNA Amplification Kit - FAQs
View additional product information for MessageAmp™ II-Bacteria RNA Amplification Kit - FAQs (AM1790)
1 product FAQs found
What is the minimum amount of input RNA to be used with the MessageAmp II-Bacteria RNA Amplification kit?
The kit can amplify as little as 10 ng of mRNA or 100 ng of total RNA.