What is the RNA input requirement when using the MessageAmp Premier RNA Amplification kit?
The RNA input recommendations is 20-500 ng total RNA, depending on the tissue or cell type.
Search
View additional product information for MessageAmp™ Premier RNA Amplification Kit - FAQs (AM1792)
1 product FAQs found