PARIS™ Kit - FAQs
PARIS™ Kit - FAQs
View additional product information for PARIS™ Kit - FAQs (AM1921)
2 product FAQs found
Can I use nuclear or cytoplasmic cell partitions with the PARIS kit?
RNA and/or protein can be isolated from either whole cells or from nuclear and cytoplasmid cell partitions.
What do you recommend for the microscale quantification of protein obtained with the PARIS Kit (Cat. No. AM1921)?
Quantitation of protein samples obtained with the PARIS kit (Cat. No. AM1921) can be assessed with standard protein assays like BCA, Bradford, or Lowry. From our Protein Assays portfolio, the following assays can be performed in microplates, require only 10 µL of sample, and show high linearity:
-Pierce 660nm Protein Assay Kit (Cat. No. 22662): working range of 50–2000 µg/mL, performed at room temperature, 5-min incubation time, and signal read at 660 nm.
-Pierce Microplate BCA Protein Assay Kit - Reducing Agent Compatible (Cat. No. 23252): working range of 125-2000 µg/mL, performed at 37 degrees C, 50 min incubation, and signal read at 562 nm.
-Pierce Rapid Gold BCA Protein Assay Kit (Cat. No. A53225): working range of 125-2000 µg/mL, performed at room temperature, 5 min incubation, and signal read at 480 nm.
To identify, compare, and choose the best protein assay for your sample type and application, you can use our interactive Protein Assay Selection Guide.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.