What is the expected yield of mRNA from total RNA?
mRNA yield is usually 1 to 3% of the total RNA. For example, if you begin with 100 µg of total RNA, you should expect a yield of 1 to 3 µg of mRNA.
Search
View additional product information for Poly(A)Purist™ MAG Kit - FAQs (AM1922)
1 product FAQs found