MagMAX™ Viral RNA Isolation Kit
Applied Biosystems™

MagMAX™ Viral RNA Isolation Kit

Ambion's MagMAX™ Viral RNA Isolation kits have been optimized for use with biological fluids and cell-free samples such as serum,Read more
Have Questions?
Catalog NumberQuantity
AM193950 preps
Catalog number AM1939
Price (USD)
394.65
Online exclusive
439.00
Save 44.35
Each
Quantity:
50 preps
Price (USD)
394.65
Online exclusive
439.00
Save 44.35
Each
Ambion's MagMAX™ Viral RNA Isolation kits have been optimized for use with biological fluids and cell-free samples such as serum, plasma, swabs, and cell culture media. RNA isolation from cell-free samples presents some unique challenges compared to RNA isolation from tissues or cultured cells. These include low viral titers, large volumes to be processed, and sample complexity. Also, the procedure must be compatible with real-time qRT-PCR. The MagMAX™ Viral RNA isolation kits address all of these issues, without the need for organic solvents or RNA precipitation, eliminating the problems often encountered with filter-based RNA isolation methods such as clogging, large elution volumes, and inconsistent yields.
• More effective recovery of viral RNA and DNA from diverse sample types
• Improved qRT-PCR results
• Ideal when working with low viral concentrations
• Improved yields and reproducibility


MagMAX™ Viral RNA Isolation Kit
The improved version of the MagMAX™ Viral RNA Isolation Kit provides better viral particle lysis and more effective recovery of both viral RNA and DNA from a variety of cell-free biological samples, including plasma, sera, and swabs. The results of these changes are improved yields and reproducibility.

The MagMAX™ Viral RNA Isolation Kit can efficiently isolate viral RNA and DNA from samples as large as 400 μl. RNA and DNA recovery is typically greater than 75%, but may vary depending on sample type. The RNA recovered with the kit is of high quality and purity and is suitable for real-time RT-PCR.

For a NVSL-validated version of this kit, please see MagMAX™ AI/ND Viral RNA Isolation Kit.

Accessory Products
Lysis/Binding Solution Concentrate, Wash Solution 1 Concentrate, Wash Solution 2 Concentrate, and Carrier RNA are components of the MagMAX™ Viral RNA Isolation Kits that may also be purchased separately.

Additional Processing Plates and Lids: we recommend 96-well plates and lids from Phenix Research Products (#MPU-8117, #MPE-8019).

Benefits of Magnetic Bead-based Isolation
Magnetic beads offer many benefits for isolating RNA from cells and viral RNA from cell-free samples. Ambion’s MagMAX™technology binds RNA more efficiently than glass fiber filter methods resulting in higher RNA yields. The RNA yields are more consistent, both from experiment to experiment, and over a broad range of sample sizes. MagMAX™magnetic bead technology eliminates filter clogging due to cellular particulates in samples. Since only a small volume of magnetic beads is needed for high efficiency binding, the bound RNA can be eluted in just 20–50 μl of nuclease free water. This makes the MagMAX™ magnetic bead technology extremely useful for concentrating RNA from large, dilute samples.
For Veterinary Use Only.
Specifications
Final Product TypeViral DNA⁄RNA
For Use With (Application)Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
High-throughput CompatibilityNot High-throughput Compatible (Manual)
No. of Reactions50
Product LineMagMAX
Purification Time45 min.
Quantity50 preps
ScaleMini
Starting Material AmountUp to 400 μl of sample
Yield10 ng
FormatTube
Isolation TechnologyMagnetic Bead
Sample TypeCell-free Biological Fluids, Liquid Samples (e.g. Serum), Plasma, Serum, Viral Samples
TargetViral DNA/RNA
Unit SizeEach
Contents & Storage
Processing Plate & Lid, Binding Solution Concentrate, Wash Solution 1 Concentrate, Wash Solution 2 Concentrate, Elution Buffer are all stored at room temperature. RNA Binding Beads are stored at 4°C and the Lysis ⁄Binding Enhancer is stored at –20°C.

Frequently asked questions (FAQs)

Is the the direct lysis approach to DNA/RNA analysis applicable for extraction of viral RNA?

Some customers have isolated RNA from viruses. Unfortunately, the protein coat could cause problems. Another option would be to use the MagMax Viral Isolation Kit.

Can I edit the manufactured KingFisher Flex protocols?

No, manufactured protocols are locked for editing, but they can be renamed using the ‘save as' function and edited as a new version.

Can I use magnetic beads from other vendors with the KingFisher Flex instrument?

For nucleic acid extraction, we recommend using our MagMAX kits and MagJET kits. Other magnetic beads should work well as long as they are 0.5 to 10 µm in size. However, you will have to write your own KingFisher protocols using the Thermo Scientific BindIt software and validate/optimize the protocol on your own.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

I am working with liquid samples with high volume and want to extract nucleic acid from them using the KingFisher Flex instrument. Can I use more volume than the protocol advises?

Yes, this can be done by sample splitting. Set the protocol so that the magnet will collect samples from multiple wells before continuing to the next step. Be sure not to overload.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

Can I scale up my reaction volume with the KingFisher Flex instrument?

Yes, reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.