What is the concentration of RNase A in the RNaseAlert Lab Test Kit (Cat. No. AM1964)?
The concentration of RNase A in the RNaseAlert Lab Test Kit is proprietary.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center