RNaseAlert™ QC System
Invitrogen™
RNaseAlert™ QC System
Ambion RNaseAlert QC System detects RNase activity in a convenient and sensitive fluorimetric assay that delivers results in real time.Read more
Have Questions?
Catalog NumberQuantity
AM1966480 Assays
Catalog number AM1966
Price (USD)
1,482.00
Each
Quantity:
480 Assays
Price (USD)
1,482.00
Each
Ambion RNaseAlert QC System detects RNase activity in a convenient and sensitive fluorimetric assay that delivers results in real time. Contain sufficient reagents for 5 × 96 high throughput assays.
• Detects as little as 3.5 x 10-7 units (∼0.5 pg) of RNase A with a fluorometer
• Simple, straightforward 30 min assay
• Monitor RNase activity in column fractions during protein purification
• Use with DNaseAlert™ QC System to quantitatively detect both RNases and DNases in the same sample
The RNaseAlert QC System uses a novel RNA substrate tagged with a fluorescent reporter molecule (fluor) on one end and a quencher on the other. In the absence of RNases, the physical proximity of the quencher dampens fluorescence from the fluor to extremely low levels. When RNases are present, however, the RNA substrate is cleaved, and the fluor and quencher are spatially separated in solution. This causes the fluor to emit a bright green signal when excited by light of the appropriate wavelength. Fluorescence can be readily detected with a filter-based or monochromator-based fluorometer. Since the fluorescence of the RNaseAlert™ Substrate increases over time when RNase activity is present, results monitored with a fluorometer can be evaluated kinetically. The sequence of the RNaseAlert Substrate has been carefully optimized to detect several RNases, including RNase A, RNase T1, RNase I, micrococcal nuclease, S1 nuclease, mung bean nuclease, and Benzonase.
Rapid and Convenient Protocol
The RNaseAlert procedure requires just minutes to set up. The lyophilized, fluorescent substrate is reconstituted in the supplied tube and incubated with the test solution for a few minutes. The results are then read on a fluorometer. Samples that noticeably fluoresce compared to the negative control are contaminated and should not be used with RNA. Nuclease-free water and RNase A are provided for use as controls.
Accessory Product:
RNaseAlert is also fully compatible with the Ambion DNaseAlert QC System (SKU# AM1970). Since the DNaseAlert Substrate contains a fluorescent tag that is spectrally distinct from RNaseAlert, the two kits can be used simultaneously with a fluorometer for quantitative real-time analyses of both RNases and DNases within a single sample.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatLyophilized
Quantity480 Assays
Reagent TypeRNase Control Reagent
Shipping ConditionDry Ice
Product LineAmbion, RNaseAlert
Unit SizeEach
Contents & Storage
Store RnaseAlert Substrate, 10X Rnase Alert Buffer, Rnase A, and TE Buffer pH 7.0 at -20°C.
Store RNaseZap™ Solution and Nuclease-free Water at room temperature.