Ambion™ RNase A, affinity purified, 1 mg/mL, 1 mg - FAQs
Ambion™ RNase A, affinity purified, 1 mg/mL, 1 mg - FAQs
View additional product information for Ambion™ RNase A, affinity purified, 1 mg/mL - FAQs (AM2271, AM2270)
1 product FAQs found
I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?
Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.