RNase A (RPA grade) 1 mg/mL - FAQs
RNase A (RPA grade) 1 mg/mL - FAQs
View additional product information for RNase A (RPA grade) 1 mg/mL - FAQs (AM2272)
1 product FAQs found
I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?
Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.