I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?
Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.
Search
View additional product information for RNase A (1 μg/mL) - FAQs (AM2274)
1 product FAQs found