RNase V1 (0.1 U/μL) - FAQs
RNase V1 (0.1 U/μL) - FAQs
View additional product information for RNase V1 (0.1 U/μL) - FAQs (AM2275)
1 product FAQs found
I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?
Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.