RNase T1 (cloned) 1,000 U/μL - FAQs
RNase T1 (cloned) 1,000 U/μL - FAQs
View additional product information for RNase T1 (cloned) 1,000 U/μL - FAQs (AM2280)
1 product FAQs found
I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?
Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.