RNase Cocktail™ Enzyme Mix, 5 x 1 mL - FAQs
RNase Cocktail™ Enzyme Mix, 5 x 1 mL - FAQs
View additional product information for RNase Cocktail™ Enzyme Mix - FAQs (AM2288, AM2286)
1 product FAQs found
I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?
Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.